missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Importin 11 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29448
This item is not returnable.
View return policy
Description
Importin 11 Polyclonal antibody specifically detects Importin 11 in Human samples. It is validated for Immunoprecipitation
Specifications
| Importin 11 | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation | |
| Unconjugated | |
| imp11, importin 11, Ran binding protein 11, Ran-binding protein 11, RANBP11, RanBP11importin-11, SLRN | |
| Between 825 and 875 | |
| 100 μg | |
| Signal Transduction | |
| 51194 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction