missing translation for 'onlineSavingsMsg'
Learn More

Importazole, MedChemExpress

Product Code. 16493258
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16493258

Brand: Medchem Express HY101091

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Importazole is a small molecule inhibitor of the nuclear transport receptor importin-β.

Importazole is a small molecule inhibitor of the nuclear transport receptor importin-β.

Specifications

Chemical Name or Material Importazole
Physical Form Solid
CAS 662163-81-7
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C20H22N4
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 33 mg/mL (103.64 mM)
SMILES CC(NC1=C2C=CC=CC2=NC(N3CCCC3)=N1)C4=CC=CC=C4
Molecular Weight (g/mol) 318.42
Formula Weight 318.42
Percent Purity 99.69%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.