missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IMPG2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564965
This item is not returnable.
View return policy
Description
Immunogen sequence: FDGGLGSGSGQ KVDLITWPWS ETSSEKSAEP LSKPWLEDDD SLLPAEIEDK KLVLVDKMDS TDQISKHSKY EHDDRSTHFP EEEPLSGPAV PIFADTAAES ASLTLPKHIS EVPGVDDYSV TKA Highest antigen sequence identity to the following orthologs - mouse 49%, rat 48%.
Chondroitin sulfate- and hyaluronan-binding proteoglycan involved in the organization of interphotoreceptor matrix; may participate in the maturation and maintenance of the light-sensitive photoreceptor outer segment. Binds heparin.
Specifications
| IMPG2 | |
| Polyclonal | |
| Unconjugated | |
| IMPG2 | |
| IMPG2; Interphotoreceptor matrix proteoglycan 2; interphotoreceptor matrix proteoglycan IPM 200; interphotoreceptor matrix proteoglycan of 200 kDa; IPM 200; IPM200; PG10.2; RP56; Rsbp; Sialoprotein associated with cones and rods proteoglycan; spacrcan; VMD5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 50939 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.10 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BZV3 | |
| IMPG2 | |
| Recombinant Human IMPG2. Recombinant protein control fragment (Product #RP-105736). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction