missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IMPG1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570669
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Human: 100%.
May interact with hyaluronan which may serve to form a basic macromolecular scaffold comprising the insoluble interphotoreceptor matrix.
Specifications
| IMPG1 | |
| Polyclonal | |
| Unconjugated | |
| IMPG1 | |
| GP147; IMPG1; interphotoreceptor matrix proteoglycan 1; Interphotoreceptor matrix proteoglycan of 150 kDa; IPM150; IPM-150; sialoprotein associated with cones and rods; SPACR; VMD4 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 3617 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q17R60 | |
| IMPG1 | |
| synthetic peptide directed towards the middle region of human IMPG1 (aa 734-783). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction