missing translation for 'onlineSavingsMsg'
Learn More

Imperialine, TRC

Product Code. 30391542
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391542 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30391542 Supplier Toronto Research Chemicals Supplier No. TRCI49250050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Imperialine
CAS 61825-98-7
Molecular Formula C27H43NO3
Quantity 50 mg
Synonym Benzo[7,8]fluoreno[2,1-b]quinolizine, cevan-6-one deriv., Kashmirine, NSC 282170Sipeimine
InChI Formula InChI=1S/C27H43NO3/c1-15-4-7-25-27(3,31)21-6-5-17-18(20(21)14-28(25)13-15)11-22-19(17)12-24(30)23-10-16(29)8-9-26(22,23)2/h15-23,25,29,31H,4-14H2,1-3H3/t15-,16-,17+,18+,19-,20-,21+,22-,23+,25-,26+,27-/m0/s1
SMILES C[C@H]1CC[C@@H]2N(C1)C[C@H]3[C@@H]4C[C@H]5[C@@H](CC(=O)[C@H]6C[C@@H](O)CC[C@]56C)[C@@H]4CC[C@H]3[C@]2(C)O
Molecular Weight (g/mol) 429.64
Formula Weight 429.3243
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.