Learn More
IMPDH2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12948-1-AP-150UL
Description
The IMPDH2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human IMPDH2. This antibody recognizes human, mouse, rat antigen. The IMPDH2 antibody has been validated for the following applications: IF, IHC, IP, WB, ELISA analysis.
Specifications
| IMPDH2 | |
| Polyclonal | |
| Unconjugated | |
| 150 μL | |
| 3615 | |
| IgG |
| Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence | |
| 260 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Rat, Mouse |