missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IMPDH2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA527519
This item is not returnable.
View return policy
Description
Recommended positive controls: HepG2, NIH-3T3, JC, BCL-1. Predicted reactivity: Mouse (100%), Rat (99%), Zebrafish (94%), Xenopus laevis (96%), Chicken (96%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes the rate-limiting enzyme in the de novo guanine nucleotide biosynthesis. It is thus involved in maintaining cellular guanine deoxy- and ribonucleotide pools needed for DNA and RNA synthesis. The encoded protein catalyzes the NAD-dependent oxidation of inosine-5'-monophosphate into xanthine-5'-monophosphate, which is then converted into guanosine-5'-monophosphate. This gene is up-regulated in some neoplasms, suggesting it may play a role in malignant transformation.
Specifications
| IMPDH2 | |
| Polyclonal | |
| Unconjugated | |
| IMPDH2 | |
| HAMAP-Rule:MF_03156}; IMP (inosine 5'-monophosphate) dehydrogenase 2; IMP (inosine monophosphate) dehydrogenase 2; IMP dehydrogenase 2; IMP dehydrogenase 2 {ECO:0000255; IMP dehydrogenase type II; IMP oxireductase 2; IMPD; IMPD 2; IMPD 2 {ECO:0000255; IMPD2; IMPDH 2; IMPDH 2 {ECO:0000255; IMPDH II; Impdh2; IMPDH-II; inosine 5' phosphate dehydrogenase 2; inosine 5-monophosphate dehydrogenase 2; inosine 5'-phosphate dehydrogenase 2; inosine monophosphate dehydrogenase 2; inosine monophosphate dehydrogenase type II; inosine-5'-monophosphate dehydrogenase 2; inosine-5'-monophosphate dehydrogenase 2 {ECO:0000255 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 23918, 3615 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 2 mg/mL | |
| PBS with 40% glycerol and 0.01% thimerosal; pH 7 | |
| P12268, P24547 | |
| IMPDH2 | |
| Recombinant protein encompassing a sequence within the center region of human IMPDH2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction