missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IMPDH1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£140.00 - £283.00
Specifications
| Antigen | IMPDH1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18395362
|
Bio-Techne
NBP3-15495-20UL |
20 μg |
£140.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18315886
|
Novus Biologicals
NBP3-15495-100UL |
100 μg |
£283.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IMPDH1 Polyclonal antibody specifically detects IMPDH1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| IMPDH1 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| DKFZp781N0678, EC 1.1.1.205, IMP (inosine 5'-monophosphate) dehydrogenase 1, IMP (inosine monophosphate) dehydrogenase 1, IMP dehydrogenase 1, IMPD, IMPD 1, IMPD1, IMPDH 1, IMPDH-I, inosine-5'-monophosphate dehydrogenase 1, LCA11RP10, retinitis pigmentosa 10 (autosomal dominant), sWSS2608 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 434-563 of human IMPDH1 (NP_899066.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Endocrinology, Neuroscience, Signal Transduction | |
| PBS, 50% glycerol, pH7.3 | |
| 3614 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title