missing translation for 'onlineSavingsMsg'
Learn More

Impact System™ Stainless Steel Sieves

Product Code. 15338313
Click to view available options
Pore Size:
125 μm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15338313

Brand: Impact Test Equipment SV614

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Quantity 1Each
Pore Size 125 μm
Type Sieve
Certifications/Compliance ISO 3310
Material Stainless Steel
Diameter (Metric) Outer 300 mm
Frame Material Stainless Steel
Mesh Material Woven Wire
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.