missing translation for 'onlineSavingsMsg'
Learn More

Impact System™ Stainless Steel Sieves

Brand:  Impact Test Equipment SV638

Code : NEW

Additional Details : Weight : 1.00000kg

Product Code. 15388313

  • £95.00 / Each
Estimated Shipment: 02-08-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Woven Wire
300 mm
1mm
ISO 3310
Stainless Steel
1Each
Sieve
Stainless Steel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers