missing translation for 'onlineSavingsMsg'
Learn More

Impact System™ Stainless Steel Sieves

Product Code. 15368313
Change view
Click to view available options
Pore Size:
355 μm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size unitSize
15368313 355 μm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15368313 Supplier Impact Test Equipment Supplier No. SV626

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Quantity 1Each
Pore Size 355 μm
Type Sieve
Certifications/Compliance ISO 3310
Material Stainless Steel
Diameter (Metric) Outer 300 mm
Frame Material Stainless Steel
Mesh Material Woven Wire
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.