Learn More
Honeywell™ Impact Pro Amplification Earmuff
Product Code. 15886825
Change view
Click to view available options
Black, NNR: 30, SNR: 33
Product Code.
15886825 Black, NNR: 30, SNR: 33
1 options
Product Code. 15886825 Supplier Honeywell™ Supplier No. 1018953
Estimated Shipment: 13-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Designed for handgun and pistol shooters looking for a high noise attenuation rating.
  • Voice amplification for communication on the range
  • Automatic 4-hour shut-off increases battery life
  • Connects to MP3 players, smart phones, and scanner radios
  • External battery compartment
  • Rubberized pressure points will prevent gunstock scratching
  • One single power and volume control knob
  • Aggressive tactical styling
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EU Directives: 89/686/EEC, EC Category PPE: Category II, Standards: EN 352-1:2002, EN 352-4:2001, EN 352-8:2008, ISO 4869-1:1990, ISO 4869-2:1994, ISO 4869-3:2007, Laboratory: PZT GmbH, An der Junkerei 48 F D-26389 Wilhelmshaven Germany, Notified Body Number: 1974
For Use With (Application) Government, law enforcement, military, noise cancellation, firearms
Noise Reduction Rating (NRR) NNR: 30 dB, SNR: 33 dB
Includes 2 AAA batteries and 3.5 audio input connection cord