missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IMPA2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA521505
This item is not returnable.
View return policy
Description
Recommended positive controls: H1299, Molt-4. Predicted reactivity: Mouse (93%), Rat (93%), Rhesus Monkey (99%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This locus encodes an inositol monophosphatase. The encoded protein catalyzes the dephosphoylration of inositol monophosphate and plays an important role in phosphatidylinositol signaling. This locus may be associated with susceptibility to bipolar disorder.
Specifications
| IMPA2 | |
| Polyclonal | |
| Unconjugated | |
| IMPA2 | |
| 2210415D20Rik; AI326924; AW259601; hypothetical protein LOC553595; IMP 2; IMP.18P; Impa2; IMPase 2; inosine monophosphatase 2; inositol (myo)-1(or 4)-monophosphatase 2; inositol monophosphatase 2; inositol monophosphatase 2 variant 1; inositol monophosphatase 2 variant 2; inositol(myo)-1(or 4)-monophosphatase 2; inositol-1(or 4)-monophosphatase 2; myo-inositol monophosphatase 2; myo-inositol monophosphatase A2; zgc:110201 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3613 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.3 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| O14732 | |
| IMPA2 | |
| Recombinant protein encompassing a sequence within the center region of human IMPA2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction