missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IMPA2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA556189

Product Code. 15744101

  • £467.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: WEECFQAAVQ LALRAGQIIR KALTEEKRVS TKTSAADLVT ETDHLVEDLI ISELRERFPS HRFIAEEAAA SGAKCVLTHS PTWIIDPI Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 91%.

This locus encodes an inositol monophosphatase. The encoded protein catalyzes the dephosphoylration of inositol monophosphate and plays an important role in phosphatidylinositol signaling. This locus may be associated with susceptibility to bipolar disorder.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IMPA2
Polyclonal
Unconjugated
IMPA2
2210415D20Rik; AI326924; AW259601; hypothetical protein LOC553595; IMP 2; IMP.18P; Impa2; IMPase 2; inosine monophosphatase 2; inositol (myo)-1(or 4)-monophosphatase 2; inositol monophosphatase 2; inositol monophosphatase 2 variant 1; inositol monophosphatase 2 variant 2; inositol(myo)-1(or 4)-monophosphatase 2; inositol-1(or 4)-monophosphatase 2; myo-inositol monophosphatase 2; myo-inositol monophosphatase A2; zgc:110201
Rabbit
Antigen affinity chromatography
RUO
3613
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
O14732
IMPA2
Recombinant protein corresponding to Human IMPA2. Recombinant protein control fragment (Product #RP-91745).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.