missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IMPA1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA576923
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
IMPA1, also known as Myo-inositol monophosphatase 1, is responsible for the procurement of inositol that is required for synthesis of phosphatidylinositol and polyphosphoinositides. IMPA1 exists as a homodimer and has been identified as the pharmacological target for lithium action in the brain. IMPA1 is the principal enzyme of the phosphatidyl inositol signaling pathway, and inhibition of inositol monophosphatase hydrolysis may underlie the anti-manic and anti-depressant actions of Li+. Studies indicate that a variation in the 277 codon coding region of the IMPA1 gene has not been observed in manic-depressive patients, therefore suggesting that polymorphisms or mutations in the noncoding regions of this gene may influence the lithium response in psychiatric patients.
Specifications
| IMPA1 | |
| Polyclonal | |
| Unconjugated | |
| Impa1 | |
| 2610002K09Rik; 2900059K10Rik; AI325909; D-galactose 1-phosphate phosphatase; IMP; IMP 1; IMPA; IMPA1; Impa1-L; IMPase 1; inositol (myo)-1(or 4)-monophosphatase 1; inositol monophosphatase 1; inositol(myo)-1(or 4)-monophosphatase 1; inositol-1(or 4)-monophosphatase 1; Lithium-sensitive myo-inositol monophosphatase A1; myo-inositol monophosphatase 1; testicular tissue protein Li 94 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3612, 55980, 83523 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| O55023, P29218, P97697 | |
| Impa1 | |
| Recombinant full length Human IMPA1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction