missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IMPA1 Antibody (2G5), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59416PE
This item is not returnable.
View return policy
Description
IMPA1 Monoclonal antibody specifically detects IMPA1 in Human samples. It is validated for Flow Cytometry
Specifications
| IMPA1 | |
| Monoclonal | |
| PE | |
| EC 3.1.3.25, IMP, IMP 1, IMPA, IMPase 1, inositol monophosphatase 1, inositol(myo)-1(or 4)-monophosphatase 1, Inositol-1(or 4)-monophosphatase 1, Lithium-sensitive myo-inositol monophosphatase A1 | |
| Recombinant human IMPA1 (1-277aa) purified from E. coli | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 3612 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry | |
| 2G5 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto