missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IMP5 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12664-1-AP-150UL
This item is not returnable.
View return policy
Description
Intramembrane-cleaving aspartic protease (I-CLiP) that may be able to cleave type II membrane signal peptides in the hydrophobic plane of the membrane.Specifications
| IMP5 | |
| Polyclonal | |
| Unconjugated | |
| SPPL2C | |
| IMP 5, IMP5, intramembrane protease 5, SPP like 2C, SPPL2c | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 162540 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.18 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8IUH8 | |
| SPPL2C | |
| IMP5 Fusion Protein Ag3345 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction