Learn More
IMP5 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12664-1-AP-20UL
 View more versions of this product
Product Code. 16850504
  • £135.00 / 20µL
Description
Intramembrane-cleaving aspartic protease (I-CLiP) that may be able to cleave type II membrane signal peptides in the hydrophobic plane of the membrane.
Specifications
Specifications
IMP5
Polyclonal
Unconjugated
SPPL2C
IMP 5, IMP5, intramembrane protease 5, SPP like 2C, SPPL2c
Rabbit
Antigen Affinity Chromatography
RUO
162540
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation, Immunocytochemistry, Immunofluorescence
0.18 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8IUH8
SPPL2C
IMP5 Fusion Protein Ag3345
20 μL