missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IMP4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566830
This item is not returnable.
View return policy
Description
Immunogen sequence: AQRMNRGRHEV GALVRACKAN GVTDLLVVHE HRGTPVGLIV SHLPFGPTAY FTLCNVVMRH DIPDLGTMSE AKPHLITHGF SSR Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
IMP4 forms a ternary complex with IMP3.
Specifications
| IMP4 | |
| Polyclonal | |
| Unconjugated | |
| Imp4 | |
| AA409888; AV031295; Brix domain-containing protein 4; BXDC4; D1Wsu40e; IMP4; IMP4 homolog, U3 small nucleolar ribonucleoprotein; IMP4, U3 small nucleolar ribonucleoprotein; IMP4, U3 small nucleolar ribonucleoprotein, homolog; IMP4, U3 small nucleolar ribonucleoprotein, homolog (yeast); LRRGT00017; RGD1308463; U3 small nucleolar ribonucleoprotein protein IMP4; U3 snoRNP protein 4 homolog; U3 snoRNP protein IMP4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 92856 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96G21 | |
| Imp4 | |
| Recombinant Human IMP4. Recombinant protein control fragment (Product #RP-107487). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction