Learn More
IMP3 Mouse anti-Human, CoraLite 555, Clone: 1E7D6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL555-66247-100UL
Description
The IMP3 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human IMP3. This antibody recognizes human, mouse antigen. The IMP3 antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™555.
Specifications
| IMP3 | |
| Monoclonal | |
| CoraLite 555 | |
| 100 μL | |
| 55272 | |
| IgG2b |
| Flow Cytometry (Intracellular Staining) | |
| 1E7D6 | |
| Mouse | |
| Primary | |
| Human, Mouse |