missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IMP3 Mouse anti-Human, Clone: 3.20E+10, Novus Biologicals™
Mouse Monoclonal Antibody
£234.00 - £378.00
Specifications
| Antigen | IMP3 |
|---|---|
| Clone | 32000000000 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, ELISA, Immunocytochemistry/Immunofluorescence 1:100 |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18204105
|
Novus Biologicals
NBP2-59475-50UL |
50 μL |
£234.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18251981
|
Novus Biologicals
NBP2-59475-100UL |
100 μL |
£378.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IMP3 Monoclonal specifically detects IMP3 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| IMP3 | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Human | |
| BRMS2mitochondrial ribosomal protein S4, C15orf12, chromosome 15 open reading frame 12, FLJ10968, IMP3, U3 small nucleolar ribonucleoprotein, homolog (yeast), MRPS4DKFZp586L0118, U3 small nucleolar ribonucleoprotein protein IMP3, U3 snoRNP protein 3 homolog, U3 snoRNP protein IMP3 | |
| IMP3 | |
| IgG1 κ | |
| Protein A purified |
| 32000000000 | |
| Western Blot 1:1000, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
| 55272 | |
| Recombinant human IMP3 (1-184aa) purified from E. coli | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title