missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IMP3 Antibody (IMP3/8985R), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-24183CL1
This item is not returnable.
View return policy
Description
IMP3 Monoclonal antibody specifically detects IMP3 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| IMP3 | |
| Monoclonal | |
| CoraFluor 1 | |
| BRMS2mitochondrial ribosomal protein S4, C15orf12, chromosome 15 open reading frame 12, FLJ10968, IMP3, U3 small nucleolar ribonucleoprotein, homolog (yeast), MRPS4DKFZp586L0118, U3 small nucleolar ribonucleoprotein protein IMP3, U3 snoRNP protein 3 homolog, U3 snoRNP protein IMP3 | |
| Recombinant fragment of human IMP3 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 55272 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| IMP3/8985R | |
| PBS | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction