missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
IMP3 Monoclonal antibody specifically detects IMP3 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | IMP3 |
| Applications | Immunocytochemistry |
| Gene Alias | BRMS2mitochondrial ribosomal protein S4, C15orf12, chromosome 15 open reading frame 12, FLJ10968, IMP3, U3 small nucleolar ribonucleoprotein, homolog (yeast), MRPS4DKFZp586L0118, U3 small nucleolar ribonucleoprotein protein IMP3, U3 snoRNP protein 3 homolog, U3 snoRNP protein IMP3 |
| Immunogen | Recombinant human IMP3 (1-184aa) purified from E. coli |
| Purification Method | Protein A purified |
| Quantity | 0.1 mL |
| Regulatory Status | RUO |
| Gene ID (Entrez) | 55272 |
| Target Species | Human |
| Isotype | IgG1 κ |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?