missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IMP2/IGF2BP2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-33728
This item is not returnable.
View return policy
Description
IMP2/IGF2BP2 Polyclonal antibody specifically detects IMP2/IGF2BP2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).
Specifications
| IMP2/IGF2BP2 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| IGF2BP2 | |
| Recombinant protein encompassing a sequence within the center region of human IGF2BP2. The exact sequence is proprietary. | |
| 100 μL | |
| Cancer, Chromatin Research, Immune System Diseases, Immunology, Transcription Factors and Regulators | |
| 10644 | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.0mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000, Immunohistochemistry-Frozen | |
| Hepatocellular carcinoma autoantigen p62, IGF-II mRNA-binding protein 2, IMP-2IGF2 mRNA-binding protein 2, IMP2VICKZ family member 2, insulin-like growth factor 2 mRNA binding protein 2, insulin-like growth factor 2 mRNA-binding protein 2, p62, VICKZ2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Tested in human. Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction