missing translation for 'onlineSavingsMsg'
Learn More

IMP Dehydrogenase 2/IMPDH2 Rabbit anti-Human, Mouse, Rat, Clone: 4X3S10, Novus Biologicals™

Rabbit Monoclonal Antibody

£150.00 - £425.00

Specifications

Antigen IMP Dehydrogenase 2/IMPDH2
Clone 4X3S10
Dilution Western Blot, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:10-1:20
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18305115
View Documents
Novus Biologicals
NBP3-16786-20UL
20 μg
£150.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18370524
View Documents
Novus Biologicals
NBP3-16786-100UL
100 μg
£425.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

IMP Dehydrogenase 2/IMPDH2 Monoclonal antibody specifically detects IMP Dehydrogenase 2/IMPDH2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications

Specifications

IMP Dehydrogenase 2/IMPDH2
Western Blot, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:10-1:20
Monoclonal
Purified
RUO
Human, Mouse, Rat
EC 1.1.1.205, IMP (inosine 5'-monophosphate) dehydrogenase 2, IMP (inosine monophosphate) dehydrogenase 2, IMP dehydrogenase 2, IMPD 2, IMPD2IMP oxireductase 2, IMPDH 2, IMPDH-II, inosine 5' phosphate dehydrogenase 2, inosine monophosphate dehydrogenase type II, inosine-5'-monophosphate dehydrogenase 2
Recombinant fusion protein containing a sequence corresponding to amino acids 100-300 of human IMP Dehydrogenase 2/IMPDH2 (P12268).
Primary
Store at -20°C. Avoid freeze-thaw cycles.
4X3S10
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Lipid and Metabolism
PBS, 0.05% BSA, 50% glycerol, pH7.3
3615
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.