missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IMP Dehydrogenase 2/IMPDH2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 4 publications
Brand: Novus Biologicals NBP1-86177
This item is not returnable.
View return policy
Description
IMP Dehydrogenase 2/IMPDH2 Polyclonal specifically detects IMP Dehydrogenase 2/IMPDH2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.
Specifications
| IMP Dehydrogenase 2/IMPDH2 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| EC 1.1.1.205, IMP (inosine 5'-monophosphate) dehydrogenase 2, IMP (inosine monophosphate) dehydrogenase 2, IMP dehydrogenase 2, IMPD 2, IMPD2IMP oxireductase 2, IMPDH 2, IMPDH-II, inosine 5' phosphate dehydrogenase 2, inosine monophosphate dehydrogenase type II, inosine-5'-monophosphate dehydrogenase 2 | |
| Rabbit | |
| 56 kDa | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 3615 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.1mg/mL | |
| Western Blot 0.04 - 0.4 ug/ml, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:200-1:500, Knockdown Validated | |
| P12268 | |
| IMPDH2 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:GFITDPVVLSPKDRVRDVFEAKARHGFCGIPITDTGRMGSRLVGIISSRDIDFLKEEEHDCFLEEIMTKREDLVVAPAGITLKEANEILQRSKKGKLPIVNEDDELVAIIARTDLKKNRDYPLASKD | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction