missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies ImmunoCult™-XF T Cell Expansion Medium, 500 mL

Product Code. 15811152
Encompass
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15811152 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15811152 Supplier STEMCELL Technologies Supplier No. 10981

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Serum-free and xeno-free medium for the expansion of human T cells

  • ImmunoCult™-XF T Cell Expansion Medium is a serum-free and xeno-free medium optimized for the in vitro culture and expansion of human T cells isolated from peripheral blood
  • Recombinant cytokines, required for the optimal growth and expansion of T cells, have not been added to ImmunoCult™-XF T Cell Expansion Medium
  • This allows users the flexibility to prepare medium that meets their requirements
  • This product is designed for cell therapy research applications following the recommendations of USP<1043> on Ancillary Materials, and we can currently work with you to qualify this reagent under an approved investigational new drug (IND) or clinical trial application (CTA)

Specifications

Cell Type T Cells
For Use With (Application) Cell Culture; Expansion
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.