missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies ImmunoCult™ Dendritic Cell Maturation Supplement
Click to view available options
Quantity:
0.5 mL
Unit Size:
0.5mL
Description
- ImmunoCult™-ACF Dendritic Cell Medium and Supplements, sold individually or as a kit, have been developed for the in vitro culture and differentiation of human monocytes into dendritic cells (DCs)
- ImmunoCult™-ACF Dendritic Cell Medium is a serum-free and animal component-free medium optimized for the in vitro culture and differentiation of human monocytes into DCs
- ImmunoCult™-ACF Dendritic Cell Differentiation Supplement contains a combination of animal component-free recombinant human cytokines formulated to support the differentiation of immature DCs from human monocytes
- The ImmunoCult™ Dendritic Cell Maturation Supplement has been formulated to support maturation of immature DCs to mature DCs
Specifications
Specifications
| Cell Type | Dendritic Cells, Monocytes |
| For Use With (Application) | Cell Culture, Differentiation |
| Quantity | 0.5 mL |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction