Learn More
Thermo Scientific™ Immuno Clear Standard Modules
Manufactured to fit standard equipment, modules provide flexible use of components with small sample sizes.
Brand: Thermo Scientific™ 6506
Code : 04
Additional Details : Weight : 5.19000kg
Description
Perform applications which are delivering well-to-well reproducibility with Thermo Scientific™ Immuno Clear Standard Modules, which feature Nunc™, Immulon™ and Microtiter™ module plates developed for ELISA techniques. Manufactured to fit standard equipment, modules provide flexible use of components with small sample sizes. They are backed by industry-leading quality standards and certifications.
Features:
- Several strip formats: Eight-well strip, 12-well strip, 16-well strip (two columns of eight wells)
- Several well shapes: flat bottom, C bottom (radius edge), U shape wells, StarWells (wells have fins to increase the surface area and enhance performance)
- Several surfaces to maximize assay performance
- Modules that remain in frame when inverted
- Stackable frame marked for lid orientation
- Polystyrene wells in polyethylene frame
- Ext. Dim.: 128 × 86mm
Choice of Surface:
- Thermo Scientific™ PolySorp™, Immulon™ 1B, and Universal Binding (UB) are for adsorption of hydrophobic molecules.
- Thermo Scientific™ Medisorp™ and Immulon 2HB are slightly hydrophilic and bind a diverse range of biomolecules.
- Thermo Scientific™ MaxiSorp™, Immulon 4 HBX, and Enhanced Binding(EB) are hydrophilic and ideal for antibody sandwich assays.
- Thermo Scientific™ MultiSorp™ are very hydrophilic and works well for glycans and water soluble proteins.
Ordering Information:
Modules are freestanding. Cat. No. 95029440 and 464394 are certified. Frames and modules are available separately. Sealing tape and strip caps are available. Color-coded modules are available on request. Contact your customer service or sales representative for further details.
Specifications
Slightly Hydrophilic | |
Standard | |
8 x 12 | |
Adsorption | |
Microplate Strip | |
96 | |
Immulon 2 HB | |
100 | |
Proteins |
No | |
Colorimetric | |
100 Boxes | |
Clear | |
Polystyrene | |
280 μL | |
Flat Bottom (F-well) | |
100 |