Learn More
IMMT Mouse, anti-Human, CoraLite Plus 488, Clone: 2D2E8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-68226-20UL
Description
The IMMT antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human IMMT. This antibody recognizes human, rat, mouse antigen. The IMMT antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| IMMT | |
| 2D2E8 | |
| CoraLite Plus 488 | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse | |
| 155 kDa | |
| IgG2b |
| Immunofluorescence | |
| 1000 μg/mL | |
| DKFZp779P1653, HMP, IMMT, Mitofilin, P87, P87/89, P89, PIG4, PIG52 | |
| 20 μL | |
| 10989 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |