Learn More
Merck Immobilon™ GO for Simple Immunodetection
Shop All Merck ProductsDescription
The Immobilon™ GO device is a hands-free immunodetection device designed for walk away and stand-alone processing of Western blots. Each device can process one mini blot. The unique design uses continuous sequential fluid convection across a flow matrix to enable hands free antibody binding and membrane wash steps performed during immunodetection of western blots.
The Immobilon™ GO device is intended for one-time use and does not require an external power or vacuum source for operation. Standard non-fat dry milk blot buffer is used throughout the Immobilon™ GO set up to allow for easy transition from traditional western blotting protocols. Western blots are processed in as little as 3 hours; however, for greater convenience blots may remain in the Immobilon™ GO device longer, even for overnight processing.
Compatible Blocking Solutions
- TBS-T with NFDM
Compatible Detection Reagents
- Immobilon Western HRP Substrates
- Commercially available detection reagents
Compatible Membranes
- PVDF (Immobilon membranes)
- Nitrocellulose
Specifications
Specifications
| Detection Method | Chemiluminescence, Fluorescence, Chromogenic |
| For Use With (Application) | Immunoblot (Western) |
| Quantity | 2/Pk. |
| Content And Storage | Room Temperature |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.