missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Immobiline™ DryStrip Gels

Product Code. 11703239 Shop All Cytiva Products
Change view
Click to view available options
pH Range:
3 to 10
3 to 11
4 to 7
6 to 11
6 to 9
Gradient:
Linear
Non-linear
Length (Metric):
7 cm
11 cm
13 cm
18 cm
24 cm
Unit Size:
Pack of 12
24 product options available for selection
Product selection table with 24 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. pH Range Gradient Length (Metric) unitSize
11703239 4 to 7 Linear 13 cm Pack of 12
11544985 3 to 10 Non-linear 18 cm Pack of 12
10753407 6 to 11 Linear 18 cm Pack of 12
11763239 6 to 11 Linear 13 cm Pack of 12
11743239 6 to 11 Linear 7 cm Pack of 12
11733249 6 to 9 Linear 24 cm Pack of 12
11713249 3 to 10 Non-linear 24 cm Pack of 12
10146414 3 to 11 Non-linear 7 cm Pack of 12
11793289 3 to 11 Non-linear 13 cm Pack of 12
11713299 3 to 11 Non-linear 24 cm Pack of 12
11524985 4 to 7 Linear 18 cm Pack of 12
11534985 3 to 10 Linear 18 cm Pack of 12
10545925 3 to 10 Linear 7 cm Pack of 12
10732637 3 to 10 Non-linear 7 cm Pack of 12
11773229 4 to 7 Linear 7 cm Pack of 12
11713239 3 to 10 Linear 13 cm Pack of 12
11723239 3 to 10 Non-linear 13 cm Pack of 12
11753239 6 to 11 Linear 11 cm Pack of 12
11723249 4 to 7 Linear 24 cm Pack of 12
10697465 3 to 10 Linear 24 cm Pack of 12
11703299 3 to 11 Non-linear 18 cm Pack of 12
10584375 3 to 11 Non-linear 11 cm Pack of 12
11723299 4 to 7 Linear 11 cm Pack of 12
11733299 3 to 10 Linear 11 cm Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 24 options available.
24 options
This item is not returnable. View return policy
Product Code. 11703239 Supplier Cytiva Supplier No. 17600113

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Represent a comprehensive selection of narrow-, medium- and wide-range IPG strips. Cytiva Life Sciences™ Immobiline™ DryStrip gels (IPG strips) are used for isoelectric focusing (IEF), run as the first dimension of 2-D electrophoresis or as a separate application.

Choose from a comprehensive range of overlapping Immobiline DryStrip gels that cover narrow (1 pH unit), medium (3 to 5 pH units), and broad (8 pH units) pH ranges. The pH gradients at the extreme ends of the pH scale are nonlinear to distribute the proteins evenly over the gel length to obtain maximum resolution.

Five narrow pH range (1 pH unit), 24-cm IPG strips deliver high resolution and protein loading capacity, which results in well separated spots with high protein content after the second-dimension separation

  • Select the broad pH 3 to 11 Non-Linear (NL) gradient Immobiline DryStrip gel for fast and efficient screening to gain a broad overview of total protein disribution
  • For more detailed studies or when using greater protein loads, choose from four new overlapping strips covering the pH 3 to 11 range
  • Higher resolution can be obtained using narrow-range (1 pH unit) Immobiline DryStrip gels
  • Use matching IPG Buffers for optimized running conditions
  • Use IPG strips directly after reswelling without further manipulation
  • Bar coded strips for convenient sample and batch tracking

  • Immobiline DryStrip gels are rehydrated with a matching IPG Buffer prior to use
  • Samples can be included directly in the rehydration solution for efficient loading of dilute samples or for loading larger amounts of protein
  • Samples can also be loaded after rehydration

TRUSTED_SUSTAINABILITY

Specifications

Description Immobiline DryStrip gels (IPG strips) are used for isoelectric focusing (IEF), run as the first dimension of 2-D electrophoresis or as a separate application.
Color Colorless
Format Precast Gel
pH Range 4 to 7
Separation Type Isoelectric Focusing
Form Dried
For Use With (Application) Industrial applications: analytical chemistry, research
Gradient Linear
Storage Requirements −20°C
Thickness (English) 0.19 in.
Thickness (Metric) 0.05 cm
Length (Metric) 13 cm
Width (English) 0.118 in.
Width (Metric) 0.3 cm
Product Type Precast Gel
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.