missing translation for 'onlineSavingsMsg'
Learn More

Grant Instruments™ Advanced Digitial Optima Immersion Thermostat

Product Code. 12787839
Change view
Click to view available options
Voltage:
120/230 V
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Voltage unitSize
12787839 120/230 V Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12787839 Supplier Grant Instruments™ Supplier No. TXF200

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High specification Optima heated circulator for sophisticated applications requiring complex programming and/or ultra precise temperature control.

  • Stability ±0.01ºC
  • Large, bright full colour display
  • Icon driven home screen via a dial and two function buttons
  • Programming/temperature profiling (TXF200 10 programs with 100 segments)
  • Model available with/without clamp (T-Clamp)
  • Low-liquid detection (float switch)
  • 5 point user calibration
  • User adjustable over temperature dial
TRUSTED_SUSTAINABILITY

Specifications

Flow Rate 23 L/min.
Max. Pressure 530 mbar
Electrical Requirements 120 V 1.5 kw 50/60 Hz, 230 V 2 kw 50/60 Hz
Voltage 120/230 V
Frequency 50/60 Hz
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.