Learn More
BD Biosciences™ IMK-Lymphocyte (CE/IVD)
Two-color direct immunofluorescence reagent kit for enumerating percentages of mature human leucocyte subsets in erythrocyte-lysed whole blood.
Brand:  BD Biosciences™ 340182
Product Code. 15855848
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Two-color direct immunofluorescence reagent kit for enumerating percentages of the following mature human leuKocyte subsets in erythrocyte-lysed whole blood (LWB): T (CD3+) lymphocytes, B (CD19+) lymphocytes, helper/inducer T (CD3+CD4+) lymphocytes, suppressor/cytotoxic T (CD3+CD8+) lymphocytes, and natural killer (NK) (CD3CD16+and/or CD56+) lymphocytes.
  • The helper/suppressor T-lymphocyte ratio (CD3+CD4+/CD3+CD8+) can also be determined.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Flow Cytometry
1 Kit
Human
Immunofluorescence and flow cytometry test
Leucogate (CD45/CD14), Control Reagent (ITC-labeled IgG1, clone X40, and PE-labeled IgG2a, clone X39), CD3/CD19, CD3/CD4, CD3/CD8, CD3/CD16+CD5, FACS Lysing Solution
CE, IVD
When stored at 2°C to 8°C, antibody reagents are stable until the expiration date shown on the label. Do not use after the expiration date. Antibody reagensst should not be frozen or exposed to direct light during storage or during incubation with cells. Keep the reagent vial dry. Incubation or centrifugation times or temperatures other than those specified can be a source of error. For optimal results, stain blood samples within 6 hours of venipuncture. Alteration in the appearance of any of the reagents, such as precipitation or discoloration, indicates instability or deterioration. In such cases, the reagent should not be used. The antibody reagents contain sodium azide as a preservative; however, care should be taken to avoid microbial contamination, which can cause erroneous results.