missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Imisopasem manganese, MedChemExpress
Description
Imisopasem manganese (M40403) is a stable non-peptidyl mimetic of manganese superoxide MnSOD.
Specifications
Specifications
| Chemical Name or Material | Imisopasem manganese |
| Physical Form | Solid |
| CAS | 218791-21-0 |
| Color | White |
| Quantity | 2 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C21H31Cl2MnN5 |
| For Use With (Application) | Cancer-programmed cell death |
| Synonym | M40403 GC4403 |
| Solubility Information | H2O : 16 mg/mL (33.38 mM; Need ultrasonic and warming) |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?