missing translation for 'onlineSavingsMsg'
Learn More

Imiquimod, TRC

Product Code. 30390964
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390964 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30390964 Supplier Toronto Research Chemicals Supplier No. TRCI475000100MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Imiquimod
CAS 99011-02-6
Molecular Formula C14 H16 N4
Quantity 100 mg
Synonym 1-(2-Methylpropyl)-1H-imidazo[4,5-c]quinolin-4-amine, 1-Isobutyl-1H-imidazo[4,5-c]quinolin-4-amine, Aldara, Beselna, Imiquimod, R 837, S 26308, TMX 101, Zyclara
InChI Formula InChI=1S/C14H16N4/c1-9(2)7-18-8-16-12-13(18)10-5-3-4-6-11(10)17-14(12)15/h3-6,8-9H,7H2,1-2H3,(H2,15,17)
SMILES CC(C)Cn1cnc2c(N)nc3ccccc3c12
IUPAC Name 1-(2-methylpropyl)imidazo[4,5-c]quinolin-4-amine
Molecular Weight (g/mol) 240.3
Formula Weight 240.1375
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.