missing translation for 'onlineSavingsMsg'
Learn More

Imiquimod, MedChemExpress

Product Code. 16428156
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16428156 100 mg 100mg
16401396 200 mg 200mg
16492046 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16428156 Supplier Medchem Express Supplier No. HYB0180
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Imiquimod (R 837), an immune response modifier, is a selective toll like receptor 7 (TLR7) agonist. Imiquimod exhibits antiviral and antitumor effects in vivo. Imiquimod can be used for the research of external genital, perianal warts, cancer and COVID-19.

Imiquimod (R 837), an immune response modifier, is a selective toll like receptor 7 (TLR7) agonist. Imiquimod exhibits antiviral and antitumor effects in vivo. Imiquimod can be used for the research of external genital, perianal warts, cancer and COVID-19.

Specifications

Chemical Name or Material Imiquimod
Physical Form Powder
CAS 99011-02-6
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C14H16N4
For Use With (Application) COVID-19-anti-virus
Synonym R 837
Solubility Information H2O : 2.64 mg/mL (10.99 mM; ultrasonic and adjust pH to 2 with HCl) ∣DMSO : 2 mg/mL (8.32 mM; ultrasonic and warming and heat to 70°C)
SMILES NC1=NC2=CC=CC=C2C3=C1N=CN3CC(C)C
Molecular Weight (g/mol) 240.3
Formula Weight 240.3
Percent Purity 98.08%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.