missing translation for 'onlineSavingsMsg'
Learn More

Imipramine Pamoate, TRC

Product Code. 30408769
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30408769 10 mg -
30405912 5 mg 5mg
30396051 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30408769 Supplier Toronto Research Chemicals Supplier No. TRCI46600810MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10075-24-8
Molecular Formula C61H64N4O6
Molecular Weight (g/mol) 949.18
Synonym 2-Naphthalenecarboxylic acid, 4,4'-methylenebis[3-hydroxy-, compd. with 10,11-dihydro-N,N-dimethyl-5H-dibenz[b,f]azepine-5-propanamine (1:2), 2-Naphthoic acid, 4,4'-methylenebis[3-hydroxy-, compd. with 5-[3-(dimethylamino)propyl]-10,11-dihydro-5H-dibenz[b,f]azepine (1:2) (8CI), 2-Naphthoic acid, 4,4'-methylenebis[3-hydroxy-, compd. with 5-[3-(dimethylamino)propyl]-10,11-dihydro-5H-dibenz[b,f]azepine (7CI), 5H-Dibenz[b,f]azepine, 5-[3-(dimethylamino)propyl]-10,11-dihydro-, 4,4'-methylenebis[3-hydroxy-2-naphthoate] (2:1) (8CI), 5H-Dibenz[b,f]azepine-5-propanamine, 10,11-dihydro-N,N-dimethyl-, 4,4'-methylenebis[3-hydroxy-2-naphthalenecarboxylate] (2:1) (9CI), Imipramine pamoate
IUPAC Name 4-[(3-carboxy-2-hydroxynaphthalen-1-yl)methyl]-3-hydroxynaphthalene-2-carboxylic acid;3-(5,6-dihydrobenzo[b][1]benzazepin-11-yl)-N,N-dimethylpropan-1-amine
SMILES CN(C)CCCN1c2ccccc2CCc3ccccc13.OC(=O)c4cc5ccccc5c(Cc6c(O)c(cc7ccccc67)C(=O)O)c4O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C23H16O6.C19H24N2/c24-20-16(14-7-3-1-5-12(14)9-18(20)22(26)27)11-17-15-8-4-2-6-13(15)10-19(21(17)25)23(28)29;1-20(2)14-7-15-21-18-10-5-3-8-16(18)12-13-17-9-4-6-11-19(17)21/h1-10,24-25H,11H2,(H,26,27)(H,28,29);3-6,8-11H,7,12-15H2,1-2H3
Formula Weight 668.2886
Chemical Name or Material Imipramine Pamoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.