missing translation for 'onlineSavingsMsg'
Learn More

Imipramine N-Oxide Hydrate, TRC

Product Code. 30390555
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390555 100 mg 100mg
30405668 10 mg -
30401488 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30390555 Supplier Toronto Research Chemicals Supplier No. TRCI466005100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1215681-42-7
Molecular Formula C19H24N2O • x(H2O)
Molecular Weight (g/mol) 296.411802
Synonym 10,11-Dihydro-N,N-dimethyl-5H-dibenz[b,f]azepine-5-propanamine N-Oxide Hydrate, 5-[3-(Dimethylamino)propyl]-10,11-dihydro-5H-dibenz[b,f]azepine 5-oxide Hydrate, Imipraminoxide Hydrate
IUPAC Name 3-(5,6-dihydrobenzo[b][1]benzazepin-11-yl)-N,N-dimethylpropan-1-amine oxide;hydrate
SMILES CN(CCCN1C2=C(C=CC=C2)CCC3=C1C=CC=C3)(C)=O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C19H24N2O.H2O/c1-21(2,22)15-7-14-20-18-10-5-3-8-16(18)12-13-17-9-4-6-11-19(17)20;/h3-6,8-11H,7,12-15H2,1-2H3;1H2
Formula Weight 314.1994
Chemical Name or Material Imipramine N-Oxide Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.