missing translation for 'onlineSavingsMsg'
Learn More

Imipramine N-b-D-Glucuronide (>85%), TRC

Product Code. 30010191
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010191 1 mg 1mg
30007160 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30010191 Supplier Toronto Research Chemicals Supplier No. TRCI4659901MG
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 165602-94-8
Molecular Formula C25H32N2O6
Molecular Weight (g/mol) 456.53
Synonym N-β-D-Glucopyranuronosyl-10,11-dihydro-N,N-dimethyl-5H-dibenz[b,f]azepine-5-propanaminium Inner Salt, Imipramine N-Glucuronide;
SMILES C[N+](C)(CCCN1C2=CC=CC=C2CCC3=CC=CC=C31)[C@H]4[C@@H]([C@H]([C@@H]([C@H](O4)C(=O)[O-])O)O)O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C25H32N2O6/c1-27(2,24-22(30)20(28)21(29)23(33-24)25(31)32)15-7-14-26-18-10-5-3-8-16(18)12-13-17-9-4-6-11-19(17)26/h3-6,8-11,20-24,28-30H,7,12-15H2,1-2H3/t20-,21-,22+,23-,24+/m0/s1
Chemical Name or Material Imipramine N-beta-D-Glucuronide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.