missing translation for 'onlineSavingsMsg'
Learn More

Imipramine Hydrochloride, TRC

Product Code. 17962915
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17962915 5 g 5g
17944195 1 g 1g
17904475 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17962915 Supplier Toronto Research Chemicals Supplier No. TRCI4659805G
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 113-52-0
Molecular Formula C19 H24 N2 . Cl H
Molecular Weight (g/mol) 316.87
Synonym Imipramine Hydrochloride, Clomipramine Hydrochloride Imp. B (EP) as Hydrochloride, Trimipramine Maleate Imp. D (EP) as Hydrochloride, 5H-Dibenz[b,f]azepine-5-propanamine, 10,11-dihydro-N,N-dimethyl-, hydrochloride (1:1), 5H-Dibenz[b,f]azepine, 5-[3-(dimethylamino)propyl]-10,11-dihydro-, monohydrochloride (8CI), 5H-Dibenz[b,f]azepine-5-propanamine, 10,11-dihydro-N,N-dimethyl-, monohydrochloride (9CI), 5-(3-Dimethylaminopropyl)-10,11-dihydro-5H-dibenz[b,f]azepine hydrochloride, Apo-Imipramine, Censtim, Censtin, Chrytemin, DIPD, Deprinol, Depsol, Dimipressin, Dyna-Zina, Efuranol, Eupramin, Feinalmin, G 22355, Imavate, Imidobenzyle, Imidol, Imilanyle, Imipramine monohydrochloride, Imiprin, Imizin, Imizine, Imizinum, Impril, Intalpram, Iramil, Irmin, Janimine, Melipramin, Melipramine hydrochloride, N-(3-Dimethylaminopropyl)iminodibenzyl hydrochloride, N-(γ-Dimethylaminopropyl)iminodibenzyl hydrochloride, Novo-pramine, Praminil, Presamine, Promiben, Pryleugan, SK-Pramine, Surplix, Teperine, Timolet, Tipramine, Tofranil, Tofranile
IUPAC Name 3-(5,6-dihydrobenzo[b][1]benzazepin-11-yl)-N,N-dimethylpropan-1-amine;hydrochloride
SMILES Cl.CN(C)CCCN1c2ccccc2CCc3ccccc13

Specifications

Quantity 5 g
InChI Formula InChI=1S/C19H24N2.ClH/c1-20(2)14-7-15-21-18-10-5-3-8-16(18)12-13-17-9-4-6-11-19(17)21;/h3-6,8-11H,7,12-15H2,1-2H3;1H
Formula Weight 316.1706
Percent Purity >95%
Chemical Name or Material Imipramine hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.