missing translation for 'onlineSavingsMsg'
Learn More

Imipramine hydrochloride

Product Code. 15485909
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15485909 25 g 25g
15475909 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15485909 Supplier Thermo Scientific Alfa Aesar Supplier No. J63723.14
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A serotonin uptake inhibitor | CAS: 113-52-0 | C19H25ClN2 | 316.873 g/mol

Imipramine hydrochloride is used as a serotonin uptake inhibitor. It mainly used in the treatment of major depression and enuresis (inability to control urination). It has also been evaluated for use in panic disorder.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Imipramine hydrochloride is used as a serotonin uptake inhibitor. It mainly used in the treatment of major depression and enuresis (inability to control urination). It has also been evaluated for use in panic disorder.

Solubility
Soluble in water

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 113-52-0
Molecular Formula C19H25ClN2
Molecular Weight (g/mol) 316.873
MDL Number MFCD00012669
InChI Key XZZXIYZZBJDEEP-UHFFFAOYSA-N
Synonym imipramine hydrochloride, imipramine hcl, tofranil, chimoreptin, chrytemin, deprinol, feinalmin, imilanyle, persamine, pertofram
PubChem CID 8228
ChEBI CHEBI:5882
IUPAC Name 3-(5,6-dihydrobenzo[b][1]benzazepin-11-yl)-N,N-dimethylpropan-1-amine;hydrochloride
SMILES CN(C)CCCN1C2=CC=CC=C2CCC3=CC=CC=C31.Cl

Specifications

Melting Point 174°C to 175°C
Color White
Quantity 25 g
Merck Index 14,4920
Solubility Information Soluble in water
Formula Weight 316.87
Physical Form Crystalline Powder
Chemical Name or Material Imipramine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.