missing translation for 'onlineSavingsMsg'
Learn More

Imipramine Hydrochloride, Mikromol™

Product Code. 17910516
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17910516 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17910516 Supplier Mikromol Supplier No. MM0077.000250
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Imipramine Hydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Imipramine hydrochloride
CAS 113-52-0
Analyte or Component Names Imipramine hydrochloride
Shipping Condition Room Temperature
Molecular Formula C19 H24 N2 . Cl H
Certifications/Compliance ISO 17034
Synonym Clomipramine Hydrochloride Imp. B (EP), Clomipramine Imp. B (EP), Trimipramine Maleate Imp. D (EP), Trimipramine Imp. D (EP), Imipramine Hydrochloride, Clomipramine Hydrochloride Imp. B (EP) as Hydrochloride, Trimipramine Maleate Imp. D (EP) as Hydrochloride, 5H-Dibenz[b,f]azepine-5-propanamine, 10,11-dihydro-N,N-dimethyl-, hydrochloride (1:1), 5H-Dibenz[b,f]azepine, 5-[3-(dimethylamino)propyl]-10,11-dihydro-, monohydrochloride (8CI), 5H-Dibenz[b,f]azepine-5-propanamine, 10,11-dihydro-N,N-dimethyl-, monohydrochloride (9CI), 5-(3-Dimethylaminopropyl)-10,11-dihydro-5H-dibenz[b,f]azepine hydrochloride, Apo-Imipramine, Censtim, Censtin, Chrytemin, DIPD, Deprinol, Depsol, Dimipressin, Dyna-Zina, Efuranol, Eupramin, Feinalmin, G 22355, Imavate, Imidobenzyle, Imidol, Imilanyle, Imipramine monohydrochloride, Imiprin, Imizin, Imizine, Imizinum, Impril, Intalpram, Iramil, Irmin, Janimine, Melipramin, Melipramine hydrochloride, N-(3-Dimethylaminopropyl)iminodibenzyl hydrochloride, N-(gamma-Dimethylaminopropyl)iminodibenzyl hydrochloride, Novo-pramine, Praminil, Presamine, Promiben, Pryleugan, SK-Pramine, Surplix, Teperine, Timolet, Tipramine, Tofranil, Tofranile, Clomipramine Hydrochloride Impurity B as Hydrochloride, Desipramine Hydrochloride Impurity A, Trimipramine Maleate Impurity D as Hydrochloride, Clomipramine Impurity B as Hydrochloride, Desipramine Impurity A, Trimipramine Impurity D as Hydrochloride
InChI Formula InChI=1S/C19H24N2.ClH/c1-20(2)14-7-15-21-18-10-5-3-8-16(18)12-13-17-9-4-6-11-19(17)21;/h3-6,8-11H,7,12-15H2,1-2H3;1H
SMILES Cl.CN(C)CCCN1c2ccccc2CCc3ccccc13
IUPAC Name 3-(5,6-dihydrobenzo[b][1]benzazepin-11-yl)-N,N-dimethylpropan-1-amine;hydrochloride
Molecular Weight (g/mol) 316.87
Quantity 250 mg
Formula Weight 316.1706 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.