missing translation for 'onlineSavingsMsg'
Learn More

Imipramine hydrochloride, MedChemExpress

Product Code. 16458776
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16458776 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16458776 Supplier Medchem Express Supplier No. HYB1490
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Imipramine hydrochloride inhibits serotonin transporter with an IC50 value of 32 nM. Imipramine hydrochloride is reported to prevent the translocation of aSMase, inhibiting MV and exosomes secretion.

Imipramine hydrochloride inhibits serotonin transporter with an IC50 value of 32 nM. Imipramine hydrochloride is reported to prevent the translocation of aSMase, inhibiting MV and exosomes secretion.

Specifications

Chemical Name or Material Imipramine hydrochloride
CAS 113-52-0
Color White
Purity Grade Notes Research
Molecular Formula C19H25ClN2
Quantity 100 mg
Solubility Information DMSO : ≥ 100 mg/mL (315.59 mM) ∣H2O : 62.5 mg/mL (197.24 mM; Need ultrasonic)
SMILES CN(C)CCCN1C2=CC=CC=C2CCC3=CC=CC=C31.[H]Cl
Molecular Weight (g/mol) 316.87
Formula Weight 316.87
Percent Purity 99.84%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.