missing translation for 'onlineSavingsMsg'
Learn More

Imipramine Hydrochloride 1.0 mg/ml in Methanol (as free base), LoGiCal

Product Code. 30460423
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30460423 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30460423 Supplier LoGiCal Supplier No. LGCAMP0077.0001
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

Imipramine Hydrochloride 1.0 mg/ml in Methanol (as free base)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Imipramine hydrochloride
CAS 113-52-0
Analyte or Component Names Imipramine hydrochloride
Shipping Condition Ice Pack (-20°C)
Molecular Formula C19 H24 N2 . Cl H
Certifications/Compliance ISO 17034
Synonym Clomipramine Hydrochloride Imp. B (EP), Clomipramine Imp. B (EP), Trimipramine Maleate Imp. D (EP), Trimipramine Imp. D (EP), Imipramine Hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride, 5-(3-Dimethylaminopropyl)-10,11-dihydro-5 H-dibenz[b,f]azepine hydrochloride
InChI Formula InChI=1 S/C19H24N2.ClH/c1-20(2)14-7-15-21-18-10-5-3-8-16(18)12-13-17-9-4-6-11-19(17)21;/h3-6,8-11 H,7,12-15H2,1-2H3;1 H
SMILES Cl.CN(C)CCCN1c2ccccc2CCc3ccccc13
IUPAC Name 3-(5,6-dihydrobenzo[b][1 ]benzazepin-11-yl)-N,N-dimethylpropan-1-amine;hydrochloride
Solvent or Matrix Methanol
Industry Type Forensics
Molecular Weight (g/mol) 316.87
Quantity 1 mL
Formula Weight 316.1706 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.