missing translation for 'onlineSavingsMsg'
Learn More

Iminostilbene, TRC

Product Code. 30401726
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30401726 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30401726 Supplier Toronto Research Chemicals Supplier No. TRCI4450001G
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Iminostilbene
CAS 256-96-2
Molecular Formula C14 H11 N
Quantity 1 g
Synonym 5H-Dibenzo[b,f]azepine, Oxcarbazepine Imp. E (EP), Carbamazepine Imp. D (EP), Iminostilbene, 2,2'-Iminostilbene, 5-Azadibenzo[a,e]cycloheptatriene, Dibenz[b,f]azepine, NSC 123458, RP 9989
InChI Formula InChI=1S/C14H11N/c1-3-7-13-11(5-1)9-10-12-6-2-4-8-14(12)15-13/h1-10,15H
SMILES N1c2ccccc2C=Cc3ccccc13
IUPAC Name 11H-benzo[b][1]benzazepine
Molecular Weight (g/mol) 193.24
Formula Weight 193.0891
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.