missing translation for 'onlineSavingsMsg'
Learn More

Iminostilbene N-Carbonyl Chloride, TRC

Product Code. 30389234
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30389234 1 g 1g
30394539 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30389234 Supplier Toronto Research Chemicals Supplier No. TRCI4450101G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5H-Dibenzo[b,f]azepine-5-carbonyl Chloride (5-Chlorocarbonyliminostilbene)
CAS 33948-22-0
Molecular Formula C15 H10 Cl N O
Quantity 1 g
Synonym 5H-Dibenzo[b,f]azepine-5-carbonyl Chloride, 5-Chlorocarbonyliminostilbene, Iminostilbene N-Carbonyl Chloride
InChI Formula InChI=1S/C15H10ClNO/c16-15(18)17-13-7-3-1-5-11(13)9-10-12-6-2-4-8-14(12)17/h1-10H
SMILES ClC(=O)N1c2ccccc2C=Cc3ccccc13
IUPAC Name benzo[b][1]benzazepine-11-carbonyl chloride
Molecular Weight (g/mol) 255.7
Formula Weight 255.0451
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.