missing translation for 'onlineSavingsMsg'
Learn More

Iminodiacetonitrile, 95%, Thermo Scientific Chemicals

£23.10 - £329.00

Chemical Identifiers

CAS 628-87-5
Molecular Formula C4H5N3
Molecular Weight (g/mol) 95.11
MDL Number MFCD00001887
InChI Key BSRDNMMLQYNQQD-UHFFFAOYSA-N
Synonym iminodiacetonitrile, 2,2'-azanediyldiacetonitrile, acetonitrile, 2,2'-iminobis, 1,1'-imidodiacetonitrile, acetonitrile, iminodi, 2,2'-iminodiacetonitrile, 2,2'-iminobisacetonitrile, bis cyanomethyl amine, di cianometil ammina, 2-cyanomethyl amino acetonitrile
PubChem CID 69413
ChEBI CHEBI:71189
IUPAC Name 2-(cyanomethylamino)acetonitrile
SMILES N#CCNCC#N
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11400068
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B23660.22
100 g
£23.10
100g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11410068
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B23660.36
500 g
£80.30
500g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11420068
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B23660.0E
2500 g
£329.00
2500g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Iminodiacetonitrile is used as a chemical intermediate. It is also used in the preparation of iminodiacetic acid. Further, it is used to study the interference of nitriles in cyanide determination.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iminodiacetonitrile is used as a chemical intermediate. It is also used in the preparation of iminodiacetic acid. Further, it is used to study the interference of nitriles in cyanide determination.

Solubility
Soluble in water, acetone and hot methanol.

Notes
Incompatible with strong acids and strong oxidizing agents.
Specifications

Chemical Identifiers

628-87-5
95.11
BSRDNMMLQYNQQD-UHFFFAOYSA-N
69413
2-(cyanomethylamino)acetonitrile
C4H5N3
MFCD00001887
iminodiacetonitrile, 2,2'-azanediyldiacetonitrile, acetonitrile, 2,2'-iminobis, 1,1'-imidodiacetonitrile, acetonitrile, iminodi, 2,2'-iminodiacetonitrile, 2,2'-iminobisacetonitrile, bis cyanomethyl amine, di cianometil ammina, 2-cyanomethyl amino acetonitrile
CHEBI:71189
N#CCNCC#N

Specifications

628-87-5
C4H5N3
773803
Soluble in water,acetone and hot methanol.
N#CCNCC#N
95.11
CHEBI:71189
95%
75°C to 79°C
MFCD00001887
iminodiacetonitrile, 2,2'-azanediyldiacetonitrile, acetonitrile, 2,2'-iminobis, 1,1'-imidodiacetonitrile, acetonitrile, iminodi, 2,2'-iminodiacetonitrile, 2,2'-iminobisacetonitrile, bis cyanomethyl amine, di cianometil ammina, 2-cyanomethyl amino acetonitrile
BSRDNMMLQYNQQD-UHFFFAOYSA-N
2-(cyanomethylamino)acetonitrile
69413
95.11
Iminodiacetonitrile
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c

H302+H312+H332

missing translation for 'einecsNumber' : 211-058-3

missing translation for 'rtecsNumber' : AM0650000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only