missing translation for 'onlineSavingsMsg'
Learn More

Iminodiacetic acid disodium salt hydrate, 97%

Product Code. 11350458
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11350458 250 g 250g
11360458 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11350458 Supplier Thermo Scientific Alfa Aesar Supplier No. 043809.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 17593-73-6 | C4H5NNa2O4 | 177.07 g/mol

Sodium iminodiacetate dibasic hydrate (Iminodiacetic acid disodium salt monohydrate) has been used in the preparation of different iminodiacetic acid-agarose supports, useful for selective adsorption of large proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17593-73-6
Molecular Formula C4H5NNa2O4
Molecular Weight (g/mol) 177.07
MDL Number MFCD00150689
InChI Key HAXVIVNBOQIMTE-UHFFFAOYSA-L
Synonym disodium iminodiacetate hydrate, iminodiacetic acid disodium salt hydrate, sodium 2,2'-azanediyldiacetate hydrate, disodium hydrate iminodiacetate, iminodiacetic acid, disodium salt monohydrate, sodium iminodiacetate dibasic hydrate, acmc-209ri5, c4h5no4.2na.h2o, disodium iminodiacetate monohydrate, sodium 2,2-azanediyldiacetate hydrate
PubChem CID 12520792
IUPAC Name disodium;2-(carboxylatomethylamino)acetate;hydrate
SMILES [Na+].[Na+].[O-]C(=O)CNCC([O-])=O

Specifications

Quantity 250 g
Merck Index 14,4917
Solubility Information Soluble in water.
Formula Weight 177.07 (Anhydrous)
Percent Purity 97%
Physical Form Crystalline
Chemical Name or Material Iminodiacetic acid disodium salt hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.