missing translation for 'onlineSavingsMsg'
Learn More

Iminodiacetic acid, 98+%

Product Code. 11404503
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11404503 500 g 500g
11494493 100 g 100g
11414503 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11404503 Supplier Thermo Scientific Alfa Aesar Supplier No. A11510.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 142-73-4 | C4H7NO4 | 133.103 g/mol

Iminoacetic acid is used as an inhibitor of bovine liver glutamate dehydrogenase. It is used as a reagent in the synthesis of poly(N-isopropylacrylamide) (PNIPAM) microgel particles for metal affinity binding of peptides. It is mainly used for modulating peptide mobility by capillary electrophoresis. It is also used in chemotherapeutic treatment for metal poisoning. It acts as a tridendate ligand and forms metal complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iminoacetic acid is used as an inhibitor of bovine liver glutamate dehydrogenase. It is used as a reagent in the synthesis of poly(N-isopropylacrylamide) (PNIPAM) microgel particles for metal affinity binding of peptides. It is mainly used for modulating peptide mobility by capillary electrophoresis. It is also used in chemotherapeutic treatment for metal poisoning. It acts as a tridendate ligand and forms metal complexes.

Solubility
Soluble in water. Insoluble in ethanol.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-73-4
Molecular Formula C4H7NO4
Molecular Weight (g/mol) 133.103
MDL Number MFCD00004280
InChI Key NBZBKCUXIYYUSX-UHFFFAOYSA-N
Synonym iminodiacetic acid, 2,2'-iminodiacetic acid, glycine, n-carboxymethyl, aminodiacetic acid, diglycocoll, diglykokoll, diglycin, iminodiethanoic acid, n-carboxymethyl glycine, 2,2'-azanediyldiacetic acid
PubChem CID 8897
ChEBI CHEBI:24786
IUPAC Name 2-(carboxymethylamino)acetic acid
SMILES C(C(=O)O)NCC(=O)O

Specifications

Melting Point ∼243°C (decomposition)
Quantity 500 g
Beilstein 878499
Merck Index 14,4917
Solubility Information Soluble in water. Insoluble in ethanol.
Formula Weight 133.1
Percent Purity ≥98%
Chemical Name or Material Iminodiacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.